Gene Transcripts

Gene Symbol
ADARB2
Entrez Gene ID
105
Gene Name
adenosine deaminase, RNA specific B2 (inactive)
Gene Aliases
ADAR3, RED2
Chromosome Location
Chr. 10: 1177313 - 1737476 on Build GRCh38
UniGene ID
Hs.586663
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018702.3 NP_061172.1 1, 2 420
GenBank mRNA AF034837.1 - 1, 2 428
BC137477.1 - 1, 2 241 9021477
BC140852.1 - 1, 2 241 9021720

View Full Product Details