Gene Transcripts

Gene Symbol
TAF3
Entrez Gene ID
83860
Gene Name
TATA-box binding protein associated factor 3
Gene Aliases
TAF140, TAFII-140, TAFII140
Chromosome Location
Chr. 10: 7818477 - 8016631 on Build GRCh38
UniGene ID
Hs.527688
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031923.3 NP_114129.1 3 1724
XM_011519741.1 XP_011518043.1 3 1748
GenBank mRNA AJ292190.1 - 3 1518
AK307521.1 - 3 1697
AK307657.1 - 3 1702
AL117661.1 - 3 1452
BC017320.1 - 3 1710
BC028077.1 - 3 1681
BC045106.1 - 3 1710
BC062352.1 - 3 1691
BC073884.1 - 3 1715

View Full Product Details