Gene Transcripts
- Gene Symbol
- CROCCP2
- Entrez Gene ID
- 84809
- Gene Name
- ciliary rootlet coiled-coil, rootletin pseudogene 2
- Gene Aliases
- CROCCL1
- Chromosome Location
- Chr. 1: 16618256 - 16630906 on Build GRCh38
- UniGene ID
- Hs.631865
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK090414.1 |
- |
11 |
3340 |
|
View Full Product Details