Gene Transcripts

Gene Symbol
C3orf79
Entrez Gene ID
152118
Gene Name
chromosome 3 open reading frame 79
Gene Aliases
Chromosome Location
Chr. 3: 153484495 - 153502697 on Build GRCh38
UniGene ID
Hs.58586
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101337.1 NP_001094807.1 1 94
GenBank mRNA AF086445.1 - 1 94

View Full Product Details