Gene Transcripts

Gene Symbol
ENDOD1
Entrez Gene ID
23052
Gene Name
endonuclease domain containing 1
Gene Aliases
Chromosome Location
Chr. 11: 95089810 - 95132651 on Build GRCh38
UniGene ID
Hs.167115
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015036.2 NP_055851.1 2 1986
GenBank mRNA AB020637.1 - 2 1937
AK026560.1 - 1 1499
BC026191.1 - 2 1953 4816880
BC071171.1 - 2 1937 5755016

View Full Product Details