Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015279.1 | NP_056094.1 | 2 | 502 | |
XM_011538078.2 | XP_011536380.1 | 2 | 444 | ||
XM_017019083.1 | XP_016874572.1 | 2 | 444 | ||
XM_017019084.1 | XP_016874573.1 | 3 | 103 | ||
XM_017019085.1 | XP_016874574.1 | 4 | 383 | ||
XM_017019086.1 | XP_016874575.1 | 4 | 388 | ||
XM_017019087.1 | XP_016874576.1 | 2 | 254 | ||
GenBank mRNA | AB023201.1 | - | 3 | 97 | |
AB449914.1 | - | 2 | 194 | ||
AK055404.1 | - | 2 | 444 | ||
AK299508.1 | - | 2 | 407 | ||
DC377016.1 | - | 2 | 503 |