Gene Transcripts
- Gene Symbol
- C2orf70
- Entrez Gene ID
- 339778
- Gene Name
- chromosome 2 open reading frame 70
- Gene Aliases
- Chromosome Location
- Chr. 2: 26562559 - 26581166 on Build GRCh38
- UniGene ID
- Hs.407083
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK091483.1 |
- |
3 |
922 |
|
View Full Product Details