Gene Transcripts
- Gene Symbol
- IGHV1OR15-1
- Entrez Gene ID
- 388077
- Gene Name
- immunoglobulin heavy variable 1/OR15-1 (non-functional)
- Gene Aliases
- IGHV1/OR15-1, IGHV1OR151
- Chromosome Location
- Chr. 15: 22160289 - 22160930 on Build GRCh38
- UniGene ID
- Hs.458446
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_135694.1 |
|
2 |
428 |
|
GenBank mRNA |
HY100667.1 |
- |
2 |
431 |
|
- Gene Symbol
- IGHV1-69
- Entrez Gene ID
- 28461
- Gene Name
- immunoglobulin heavy variable 1-69
- Gene Aliases
- IGHV1-E, IGHV169, IGHV1E
- Chromosome Location
- Chr. 14: 106714682 - 106715120 on Build GRCh38
- UniGene ID
- Hs.704366
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC070333.1 |
- |
2 |
424 |
|
- Gene Symbol
- LOC102724760
- Entrez Gene ID
- 102724760
- Gene Name
- immunoglobulin heavy variable 1/OR15-1 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 15: 21717666 - 21718307 on Build GRCh38
- UniGene ID
- Hs.458446
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_135695.1 |
|
2 |
428 |
|
GenBank mRNA |
DN998389.1 |
- |
2 |
408 |
|
View Full Product Details