Gene Transcripts

Gene Symbol
SLC50A1
Entrez Gene ID
55974
Gene Name
solute carrier family 50 member 1
Gene Aliases
HsSWEET1, RAG1AP1, SCP, SWEET1, slv
Chromosome Location
Chr. 1: 155135253 - 155138858 on Build GRCh38
UniGene ID
Hs.292154
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122837.1 NP_001116309.1 5 614
NM_001122839.1 NP_001116311.1 5 576
NM_001287586.1 NP_001274515.1 5 641
NM_001287587.1 NP_001274516.1 6 758
NM_001287588.1 NP_001274517.1 5 632
NM_001287589.1 NP_001274518.1 4 452
NM_001287590.1 NP_001274519.1 5 771
NM_001287591.1 NP_001274520.1 6 795
NM_001287592.1 NP_001274521.1 4 527
NM_018845.3 NP_061333.2 6 738
XM_005245339.4 XP_005245396.1 6 924
XM_005245340.3 XP_005245397.1 6 1005
XM_005245341.4 XP_005245398.1 6 956
XM_005245342.3 XP_005245399.1 5 564
XM_006711453.2 XP_006711516.1 6 756
GenBank mRNA AF126023.1 - 6 698
AF126024.1 - 5 563
AK289551.1 - 6 739
BC005943.1 - 6 682 4076256
BC009621.1 - 5 538 3896154
BC010869.1 - 5 529
BC012378.1 - 5 563
BE958344.1 - 594
BI195885.1 - 5 774
BX093804.1 - 4 444
CR457148.1 - 6 638
X84712.1 - 4 323

View Full Product Details