Gene Transcripts
- Gene Symbol
- NBPF10
- Entrez Gene ID
- 100132406
- Gene Name
- neuroblastoma breakpoint family member 10
- Gene Aliases
- AB6, AG1, NBPF9
- Chromosome Location
- Chr. 1: 146066318 - 146144804 on Build GRCh38
- UniGene ID
- Hs.714127
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX538005.1 |
- |
2 |
373 |
|
View Full Product Details