Gene Transcripts

Gene Symbol
TMPRSS11BNL
Entrez Gene ID
401136
Gene Name
TMPRSS11B N-terminal like, pseudogene
Gene Aliases
Chromosome Location
Chr. 4: 68184129 - 68218080 on Build GRCh38
UniGene ID
Hs.652046
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK123556.1 - 4 912

View Full Product Details