Gene Transcripts

Gene Symbol
SNX30
Entrez Gene ID
401548
Gene Name
sorting nexin family member 30
Gene Aliases
ATG24A
Chromosome Location
Chr. 9: 112750817 - 112875847 on Build GRCh38
UniGene ID
Hs.522350
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012994.1 NP_001013012.1 9 1551
GenBank mRNA AK127013.1 - 8 1255
AK128710.1 - 4 647

View Full Product Details