Gene Transcripts
- Gene Symbol
- TOLLIP
- Entrez Gene ID
- 54472
- Gene Name
- toll interacting protein
- Gene Aliases
- IL-1RAcPIP
- Chromosome Location
- Chr. 11: 1274368 - 1309666 on Build GRCh38
- UniGene ID
- Hs.368527
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318514.1 |
NP_001305443.1
|
2, 3 |
365 |
|
NM_019009.3 |
NP_061882.2
|
2, 3 |
355 |
|
GenBank mRNA |
AB209541.1 |
- |
2, 3 |
296 |
|
AJ242972.1 |
- |
2, 3 |
301 |
|
AK022871.1 |
- |
2, 3 |
262 |
|
AK127123.1 |
- |
2, 3 |
318 |
|
AK315592.1 |
- |
2, 3 |
316 |
|
AL136835.1 |
- |
2, 3 |
302 |
|
AY730683.1 |
- |
2, 3 |
191 |
|
BC004420.1 |
- |
2, 3 |
315 |
3356339 |
BC012057.1 |
- |
2, 3 |
280 |
4647828 |
BC018272.1 |
- |
2, 3 |
272 |
3452585 |
CR533477.1 |
- |
2, 3 |
191 |
|
DA227912.1 |
- |
2, 3 |
365 |
|
DB039994.1 |
- |
2, 3 |
356 |
|
HY031351.1 |
- |
2, 3 |
358 |
|
View Full Product Details