Gene Transcripts

Gene Symbol
SLC22A20
Entrez Gene ID
440044
Gene Name
solute carrier family 22 member 20
Gene Aliases
Oat6
Chromosome Location
Chr. 11: 65213840 - 65243108 on Build GRCh38
UniGene ID
Hs.532372
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_033396.1 10 1955
XM_017017775.1 XP_016873264.1 7 1762
GenBank mRNA AK094565.1 - 4 1474

View Full Product Details