Gene Transcripts

Gene Symbol
CDIPT-AS1
Entrez Gene ID
440356
Gene Name
CDIPT antisense RNA 1 (head to head)
Gene Aliases
Chromosome Location
Chr. 16: 29863683 - 29868053 on Build GRCh38
UniGene ID
Hs.448823
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015396.1 6 721
NR_024370.1 6 598
GenBank mRNA AK131031.1 - 6 721
BM006535.1 - 6 600

View Full Product Details