Gene Transcripts

Gene Symbol
LIN37
Entrez Gene ID
55957
Gene Name
lin-37 DREAM MuvB core complex component
Gene Aliases
F25965, ZK418.4, lin-37
Chromosome Location
Chr. 19: 35748361 - 35754519 on Build GRCh38
UniGene ID
Hs.529100
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019104.2 NP_061977.1 7 825
GenBank mRNA AF162447.1 - 2 189
AK293116.1 - 7 590
BC009071.1 - 7 544 3872616

View Full Product Details