Gene Transcripts

Gene Symbol
APBB1IP
Entrez Gene ID
54518
Gene Name
amyloid beta precursor protein binding family B member 1 interacting protein
Gene Aliases
INAG1, PREL1, RARP1, RIAM
Chromosome Location
Chr. 10: 26438324 - 26567879 on Build GRCh38
UniGene ID
Hs.310421
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019043.3 NP_061916.3 6 842
XM_006717451.2 XP_006717514.1 5 540
XM_011519514.2 XP_011517816.1 6 855
GenBank mRNA AB085852.1 - 4 521
AK000812.1 - 6 801
AK303597.1 - 6 1402
AK308916.1 - 6 823
AY152730.1 - 5 608
BC054516.1 - 5 620 5172946

View Full Product Details