Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031923.3 | NP_114129.1 | 2 | 548 | |
XM_011519741.1 | XP_011518043.1 | 2 | 575 | ||
GenBank mRNA | AJ292190.1 | - | 2 | 342 | |
AK307521.1 | - | 2 | 524 | ||
AK307657.1 | - | 2 | 526 | ||
AL117661.1 | - | 2 | 276 | ||
BC017320.1 | - | 2 | 534 | ||
BC028077.1 | - | 2 | 505 | ||
BC045106.1 | - | 2 | 534 | ||
BC062352.1 | - | 2 | 515 | ||
BC073884.1 | - | 2 | 539 | ||
BC105593.1 | - | 2 | 552 |