Gene Transcripts

Gene Symbol
BBIP1
Entrez Gene ID
92482
Gene Name
BBSome interacting protein 1
Gene Aliases
BBIP10, BBS18, NCRNA00081, bA348N5.3
Chromosome Location
Chr. 10: 110898730 - 110919366 on Build GRCh38
UniGene ID
Hs.232543
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195304.1 NP_001182233.1 5 1013
NM_001195305.1 NP_001182234.1 4 856
NM_001195306.1 NP_001182235.1 4 702
NM_001195307.1 NP_001182236.1 3 781
NM_001243783.1 NP_001230712.1 3 763
GenBank mRNA AK025724.1 - 4 676
AK307126.1 - 4 702
AK307314.1 - 5 889
BC105804.1 - 5 852
BI552279.1 - 3 595

View Full Product Details