Gene Transcripts

Gene Symbol
UNC45A
Entrez Gene ID
55898
Gene Name
unc-45 myosin chaperone A
Gene Aliases
GC-UNC45, GCUNC-45, GCUNC45, IRO039700, SMAP-1, SMAP1, UNC-45A
Chromosome Location
Chr. 15: 90929980 - 90954093 on Build GRCh38
UniGene ID
Hs.389461
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039675.1 NP_001034764.1 7, 8 1218
NM_001323619.1 NP_001310548.1 4, 5 477
NM_001323620.1 NP_001310549.1 4, 5 1015
NM_001323621.1 NP_001310550.1 4, 5 822
NM_018671.4 NP_061141.2 4, 5 867
XM_005254963.3 XP_005255020.1 4, 5 587
XM_011521779.2 XP_011520081.1 7, 8 1463
XM_011521780.2 XP_011520082.1 3, 4 369
XM_017022407.1 XP_016877896.1 4, 5 587
GenBank mRNA AB014729.1 - 4, 5 409
AB014736.1 - 4, 5 451
AK000905.1 - 3, 4 745
AK125721.1 - 7, 8 1218
AK291622.1 - 4, 5 422
AK293538.1 - 4, 5 591
AK296964.1 - 4, 5 444
AK302979.1 - 4, 5 484
BC006214.1 - 4, 5 450 3051384
BC037992.1 - 4, 5 450 5534906
BC045635.1 - 7, 8 1218 5297091

View Full Product Details