Gene Transcripts

Gene Symbol
SSX2
Entrez Gene ID
6757
Gene Name
SSX family member 2
Gene Aliases
CT5.2, CT5.2A, HD21, HOM-MEL-40, SSX
Chromosome Location
Chr. X: 52696896 - 52707227 on Build GRCh38
UniGene ID
Hs.289105
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278697.1 NP_001265626.1 4 365
NM_003147.5 NP_003138.3 4 365
NM_175698.2 NP_783629.1 4 365
GenBank mRNA BC002818.2 - 4 299
BC007343.2 - 4 299 3615133
BC016957.1 - 4 299 3916128
BC069313.1 - 3 229 7262129
BC071827.1 - 4 299 4641362
BC103863.2 - 4 367 40003337
BG104299.1 - 4 337
BI560469.1 - 4 372
JX014236.1 - 3 229
X86175.1 - 4 320
Z49105.1 - 4 327

Gene Symbol
SSX2B
Entrez Gene ID
727837
Gene Name
SSX family member 2B
Gene Aliases
CT5.2, CT5.2b, HOM-MEL-40, SSX
Chromosome Location
Chr. X: 52751204 - 52761536 on Build GRCh38
UniGene ID
Hs.289105
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164417.2 NP_001157889.1 4 364
NM_001278701.1 NP_001265630.1 4 364
NM_001278702.1 NP_001265631.1 4 364
GenBank mRNA BE391023.1 - 4 318
CB158955.1 - 4 281

View Full Product Details