Gene Transcripts

Gene Symbol
LCE2A
Entrez Gene ID
353139
Gene Name
late cornified envelope 2A
Gene Aliases
LEP9
Chromosome Location
Chr. 1: 152698364 - 152699442 on Build GRCh38
UniGene ID
Hs.334054
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178428.3 NP_848515.1 2 386
GenBank mRNA BC119707.2 - 2 386
BC127647.1 - 2 386
BG217525.1 - 1 360

View Full Product Details