Gene Transcripts
- Gene Symbol
- KRTAP21-1
- Entrez Gene ID
- 337977
- Gene Name
- keratin associated protein 21-1
- Gene Aliases
- KAP21.1
- Chromosome Location
- Chr. 21: 30755139 - 30755378 on Build GRCh38
- UniGene ID
- Hs.567837
- Species
- Homo sapiens
- Gene Symbol
- LOC101927016
- Entrez Gene ID
- 101927016
- Gene Name
- keratin-associated protein 21-1-like
- Gene Aliases
- Chromosome Location
- Chr. 13: 63832066 - 63833720 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- LOC647264
- Entrez Gene ID
- 647264
- Gene Name
- keratin-associated protein 6-2
- Gene Aliases
- Chromosome Location
- Chr. 13: 63746102 - 63747788 on Build GRCh38
- UniGene ID
- Hs.714690
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC157826.1 |
- |
|
79 |
|
BC171812.1 |
- |
|
79 |
9054466 |
View Full Product Details