Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|---|
RefSeq | NR_015356.2 | 3 | 321 | |||
NR_033664.1 | 2 | 250 | ||||
NR_033665.1 | 2 | 250 | ||||
GenBank mRNA | AY312364.1 | - | 3 | 324 | ||
AY312365.1 | - | LIT3317 AAA1 |
3 | 319 | ||
AY312366.1 | - | LIT3326 AAA1 |
3 | 215 | ||
AY312367.1 | - | LIT3327 AAA1 |
2 | 250 | ||
AY312368.1 | - | LIT3328 AAA1 |
2 | 250 | ||
AY312369.1 | - | LIT3332 AAA1 |
2 | 250 | ||
AY312370.1 | - | LIT3333 AAA1 |
2 | 135 | ||
AY312371.1 | - | LIT3334 AAA1 |
2 | 250 | ||
AY312372.1 | - | LIT3335 AAA1 |
2 | 250 | ||
AY312373.1 | - | LIT3331 AAA1 |
3 | 215 | ||
AY321515.1 | - | 2 | 131 | |||
AY321516.1 | - | 2 | 131 | |||
BC031961.1 | - | 3 | 302 | |||
BC130477.1 | - | 2 | 135 | |||
U85653.1 | - | 2 | 213 |