Gene Transcripts

Gene Symbol
OR13C2
Entrez Gene ID
392376
Gene Name
olfactory receptor family 13 subfamily C member 2
Gene Aliases
OR37K
Chromosome Location
Chr. 9: 104604671 - 104605627 on Build GRCh38
UniGene ID
Hs.553789
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004481.1 NP_001004481.1 1 588
GenBank mRNA BC136824.1 - 1 631
BC136826.1 - 1 631 9020817

View Full Product Details