Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001012758.2 | NP_001012776.1 | 7 | 835 | |
XM_011509260.2 | XP_011507562.1 | 5 | 1273 | ||
GenBank mRNA | AA287243.1 | - | 2 | 167 | |
AK091752.1 | - | 3 | 2094 | ||
AK298083.1 | - | 5 | 1367 | ||
BC046352.1 | - | 5 | 478 |