Gene Transcripts
- Gene Symbol
- TBC1D3P2
- Entrez Gene ID
- 440452
- Gene Name
- TBC1 domain family member 3 pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 17: 62264705 - 62275655 on Build GRCh38
- UniGene ID
- Hs.448884
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027486.1 |
|
8 |
574 |
|
GenBank mRNA |
AK301746.1 |
- |
8 |
575 |
|
BC058890.1 |
- |
8 |
599 |
|
View Full Product Details