Gene Transcripts

Gene Symbol
ODF3B
Entrez Gene ID
440836
Gene Name
outer dense fiber of sperm tails 3B
Gene Aliases
ODF3L3
Chromosome Location
Chr. 22: 50530409 - 50532579 on Build GRCh38
UniGene ID
Hs.531314
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014440.3 NP_001014440.2 7 931
GenBank mRNA BC127934.1 - 1 1667
BC127935.1 - 6 760
S72487.1 - 4 980

View Full Product Details