Gene Transcripts

Gene Symbol
SPRR2E
Entrez Gene ID
6704
Gene Name
small proline rich protein 2E
Gene Aliases
Chromosome Location
Chr. 1: 153093135 - 153094528 on Build GRCh38
UniGene ID
Hs.568518
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024209.3 NP_001019380.2 2 571
GenBank mRNA BC058018.1 - 2 586
M20030.1 - 2 570

View Full Product Details