Gene Transcripts
- Gene Symbol
- H1FX-AS1
- Entrez Gene ID
- 339942
- Gene Name
- H1FX antisense RNA 1
- Gene Aliases
- C3orf47
- Chromosome Location
- Chr. 3: 129316271 - 129324569 on Build GRCh38
- UniGene ID
- Hs.450096
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026991.1 |
|
4 |
1676 |
|
GenBank mRNA |
BC093941.1 |
- |
3 |
339 |
|
BC101739.1 |
- |
3 |
339 |
|
View Full Product Details