Gene Transcripts
- Gene Symbol
- TRIM16L
- Entrez Gene ID
- 147166
- Gene Name
- tripartite motif containing 16-like
- Gene Aliases
- TRIM70
- Chromosome Location
- Chr. 17: 18697998 - 18736119 on Build GRCh38
- UniGene ID
- Hs.164324
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001037330.1 |
NP_001032407.1
|
4, 5 |
747 |
|
XM_005256479.4 |
XP_005256536.1
|
7, 8 |
921 |
|
XM_005256480.4 |
XP_005256537.1
|
8, 9 |
1025 |
|
XM_005256481.4 |
XP_005256538.1
|
10, 11 |
1274 |
|
XM_005256483.4 |
XP_005256540.1
|
9, 10 |
1166 |
|
XM_011523689.2 |
XP_011521991.1
|
5, 6 |
626 |
|
XM_011523690.2 |
XP_011521992.1
|
11, 12 |
1336 |
|
XM_017024245.1 |
XP_016879734.1
|
12, 13 |
1636 |
|
XM_017024246.1 |
XP_016879735.1
|
11, 12 |
1498 |
|
XM_017024247.1 |
XP_016879736.1
|
12, 13 |
1639 |
|
XM_017024248.1 |
XP_016879737.1
|
11, 12 |
1528 |
|
XM_017024249.1 |
XP_016879738.1
|
10, 11 |
1390 |
|
XM_017024250.1 |
XP_016879739.1
|
10, 11 |
1391 |
|
XM_017024251.1 |
XP_016879740.1
|
10, 11 |
1228 |
|
XM_017024252.1 |
XP_016879741.1
|
5, 6 |
882 |
|
GenBank mRNA |
AK298836.1 |
- |
4, 5 |
692 |
|
AK301647.1 |
- |
4, 5 |
813 |
|
AK316368.1 |
- |
5, 6 |
781 |
|
BC127827.1 |
- |
9, 10 |
1096 |
40131713 |
BC128591.1 |
- |
7, 8 |
896 |
40127675 |
BC146902.1 |
- |
4, 5 |
650 |
9056830 |
BC146907.1 |
- |
4, 5 |
650 |
9056835 |
DQ232882.1 |
- |
4, 5 |
747 |
|
View Full Product Details