Gene Transcripts

Gene Symbol
TMEM75
Entrez Gene ID
641384
Gene Name
transmembrane protein 75
Gene Aliases
Chromosome Location
Chr. 8: 127946559 - 127948723 on Build GRCh38
UniGene ID
Hs.629674
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_103558.1 1 1443
GenBank mRNA AK093424.1 - 1 1443
AX748117.1 - 1 1443
BC137383.1 - 1 1065 9021381
BC137385.1 - 1 1065 9021383

View Full Product Details