Gene Transcripts
- Gene Symbol
- LRRC37A5P
- Entrez Gene ID
- 652972
- Gene Name
- leucine rich repeat containing 37 member A5, pseudogene
- Gene Aliases
- C9orf29
- Chromosome Location
- Chr. 9: 111602831 - 111613553 on Build GRCh38
- UniGene ID
- Hs.551134
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_034087.1 |
|
3 |
1044 |
|
GenBank mRNA |
BC031236.1 |
- |
3 |
1046 |
|
BC040897.1 |
- |
3 |
1509 |
|
View Full Product Details