Gene Transcripts
- Gene Symbol
- RNF20
- Entrez Gene ID
- 56254
- Gene Name
- ring finger protein 20
- Gene Aliases
- BRE1, BRE1A, hBRE1
- Chromosome Location
- Chr. 9: 101533849 - 101563344 on Build GRCh38
- UniGene ID
- Hs.729085
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019592.6 |
NP_062538.5
|
2, 3 |
232 |
|
XM_011518862.1 |
XP_011517164.1
|
2, 3 |
509 |
|
GenBank mRNA |
AF265230.1 |
- |
2, 3 |
230 |
|
AK000389.1 |
- |
1, 2 |
123 |
|
AK000697.1 |
- |
1, 2 |
123 |
|
AK022532.1 |
- |
2, 3 |
228 |
|
AK314401.1 |
- |
2, 3 |
187 |
|
BC016691.1 |
- |
3, 4 |
187 |
|
BC063115.1 |
- |
2, 3 |
213 |
|
BC110584.1 |
- |
2, 3 |
225 |
40009044 |
BC110585.1 |
- |
2, 3 |
225 |
40009047 |
BC152309.1 |
- |
2, 3 |
225 |
40009034 |
DB273543.1 |
- |
2, 3 |
232 |
|
View Full Product Details