Gene Transcripts

Gene Symbol
Slc46a3
Entrez Gene ID
288454
Gene Name
solute carrier family 46, member 3
Gene Aliases
RGD1307594
Chromosome Location
Chr. 12: 8725480 - 8741944 on Build Rnor_6.0
UniGene ID
Rn.16498
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024968.1 NP_001020139.1 6 2117
XM_006248833.2 XP_006248895.1 6 1945
GenBank mRNA BC091179.1 - 6 2117
FQ218873.1 - 4 827

View Full Product Details