Gene Transcripts

Gene Symbol
Tm9sf4
Entrez Gene ID
296279
Gene Name
transmembrane 9 superfamily protein member 4
Gene Aliases
Chromosome Location
Chr. 3: 148635807 - 148689961 on Build Rnor_6.0
UniGene ID
Rn.228942
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025649.1 NP_001020820.1 4, 5 615
XM_006235284.2 XP_006235346.1 4, 5 633
GenBank mRNA BC099133.1 - 4, 5 615

View Full Product Details