Gene Transcripts

Gene Symbol
Defa9
Entrez Gene ID
498658
Gene Name
defensin alpha 9
Gene Aliases
Chromosome Location
Chr. 16: 75465710 - 75466604 on Build Rnor_6.0
UniGene ID
Rn.125326
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025763.1 NP_001020934.1 2 228
GenBank mRNA AY623753.1 - 2 228

View Full Product Details