Gene Transcripts

Gene Symbol
Bckdha
Entrez Gene ID
25244
Gene Name
branched chain ketoacid dehydrogenase E1, alpha polypeptide
Gene Aliases
BCKDA, E1a
Chromosome Location
Chr. 1: 82423291 - 82452057 on Build Rnor_6.0
UniGene ID
Rn.49145
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012782.1 NP_036914.1 9 1335
GenBank mRNA BC089915.1 - 7 982
BC161819.1 - 9 1340 9099948
J02827.1 - 9 1319

View Full Product Details