Gene Transcripts

Gene Symbol
Sec62
Entrez Gene ID
294912
Gene Name
SEC62 homolog (S. cerevisiae)
Gene Aliases
Ab2-292, Tloc1
Chromosome Location
Chr. 2: 116188722 - 116225510 on Build Rnor_6.0
UniGene ID
Rn.220192
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034129.1 NP_001029301.1 6 779
XM_006232224.2 XP_006232286.1 2 361
GenBank mRNA AY325207.1 - 6 779
FQ220436.1 - 2 138

View Full Product Details