Gene Transcripts

Gene Symbol
Efhd2
Entrez Gene ID
298609
Gene Name
EF-hand domain family, member D2
Gene Aliases
Chromosome Location
Chr. 5: 160407777 - 160423811 on Build Rnor_6.0
UniGene ID
Rn.92713
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031648.1 NP_001026818.1 4 878
GenBank mRNA BC098936.1 - 4 878

View Full Product Details