Gene Transcripts

Gene Symbol
Eif3l
Entrez Gene ID
300069
Gene Name
eukaryotic translation initiation factor 3, subunit L
Gene Aliases
Eif3s6ip
Chromosome Location
Chr. 7: 120320547 - 120331596 on Build Rnor_6.0
UniGene ID
Rn.228634
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034134.1 NP_001029306.1 6 1392
GenBank mRNA BC100264.1 - 6 1392
FQ209474.1 - 1416

View Full Product Details