Gene Transcripts

Gene Symbol
Srebf2
Entrez Gene ID
300095
Gene Name
sterol regulatory element binding transcription factor 2
Gene Aliases
SREBP-2, SREBP2, Srebf2_retired
Chromosome Location
Chr. 7: 123381082 - 123438605 on Build Rnor_6.0
UniGene ID
Rn.41063
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033694.1 NP_001028866.1 8 1558
XM_006242079.2 XP_006242141.1 8 1913
GenBank mRNA BC101902.1 - 8 1558

View Full Product Details