Gene Transcripts

Gene Symbol
Zcrb1
Entrez Gene ID
362990
Gene Name
zinc finger CCHC-type and RNA binding motif 1
Gene Aliases
RGD1309851
Chromosome Location
Chr. 7: 134588902 - 134602239 on Build Rnor_6.0
UniGene ID
Rn.136969
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034940.1 NP_001030112.1 8 904
GenBank mRNA BC099747.1 - 8 904

View Full Product Details