Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020243.4 | NP_064628.1 | 4 | 1002 | |
GenBank mRNA | AB041906.1 | - | 4 | 985 | |
AF163256.1 | - | 4 | 987 | ||
AK024731.1 | - | 4 | 995 | ||
BC009363.2 | - | 4 | 974 | 4302531 | |
BQ961547.1 | - | 1 | 342 |