Gene Transcripts

Gene Symbol
Acsf2
Entrez Gene ID
619561
Gene Name
acyl-CoA synthetase family member 2
Gene Aliases
Chromosome Location
Chr. 10: 82284719 - 82326771 on Build Rnor_6.0
UniGene ID
Rn.146366
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034951.1 NP_001030123.1 9 1211
GenBank mRNA BC099826.1 - 9 1211

View Full Product Details