Gene Transcripts

Gene Symbol
Cyp4a12b
Entrez Gene ID
13118
Gene Name
cytochrome P450, family 4, subfamily a, polypeptide 12B
Gene Aliases
BC060945, Cyp4a12
Chromosome Location
Chr. 4: 115411624 - 115439034 on Build GRCm38
UniGene ID
Mm.448292
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172306.2 NP_758510.2 9, 10 1283
GenBank mRNA AK098123.1 - 9, 10 1264
BC060945.1 - 9, 10 1264 30304458

View Full Product Details