Gene Transcripts

Gene Symbol
Olfr97
Entrez Gene ID
258505
Gene Name
olfactory receptor 97
Gene Aliases
MOR156-2
Chromosome Location
Chr. 17: 37231436 - 37232368 on Build GRCm38
UniGene ID
Mm.353371
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146512.1 NP_666723.1 1 581
GenBank mRNA BC134363.1 - 585

View Full Product Details