Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020195.2 | NP_064580.2 | 4, 5 | 362 | |
XM_011536975.1 | XP_011535277.1 | 4, 5 | 406 | ||
GenBank mRNA | AF226050.1 | - | 3, 4 | 636 | |
BC000989.2 | - | 4, 5 | 362 | 2900140 | |
BC008566.1 | - | 3, 4 | 534 | ||
BI256676.1 | - | 2, 3 | 225 | ||
DT220226.1 | - | 4, 5 | 366 |