Gene Transcripts

Gene Symbol
Mettl7a2
Entrez Gene ID
393082
Gene Name
methyltransferase like 7A2
Gene Aliases
Gm921, UbiE2, Ubie
Chromosome Location
Chr. 15: 100353200 - 100361819 on Build GRCm38
UniGene ID
Mm.268451
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199477.2 NP_955771.2 2 675
GenBank mRNA AB099518.1 - 2 712
AB253944.1 - 2 666
AK143923.1 - 2 676
BC087869.1 - 2 675 30304748
BC090971.1 - 2 675 30314995

Gene Symbol
Mettl7a3
Entrez Gene ID
668178
Gene Name
methyltransferase like 7A3
Gene Aliases
ENSMUSG00000058057, Gm10035, Ubie3
Chromosome Location
Chr. 15: 100334801 - 100347682 on Build GRCm38
UniGene ID
Mm.425943
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081471.1 NP_001074940.1 2 666
GenBank mRNA AB253945.1 - 2 666

View Full Product Details