Gene Transcripts

Gene Symbol
RGMA
Entrez Gene ID
56963
Gene Name
repulsive guidance molecule family member a
Gene Aliases
RGM
Chromosome Location
Chr. 15: 93043406 - 93089214 on Build GRCh38
UniGene ID
Hs.271277
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166283.1 NP_001159755.1 4 1157
NM_001166286.1 NP_001159758.1 3 1017
NM_001166287.1 NP_001159759.1 4 1131
NM_001166288.1 NP_001159760.1 4 1015
NM_001166289.1 NP_001159761.1 4 1035
NM_020211.2 NP_064596.2 4 1184
GenBank mRNA AK074910.1 - 4 1174
AK074966.1 - 3 991
AK074980.1 - 4 951
AK125047.1 - 1 1544
AK125204.1 - 5 1140
AK291365.1 - 3 814
AK299363.1 - 4 1033
AK309814.1 - 5 1397
AL136826.1 - 4 998
AL390083.1 - 3 940
BC015886.1 - 3 1008
BC127826.1 - 1 165
BC140838.1 - 4 962 9021706
BC151132.1 - 4 962 9057032

View Full Product Details